missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Life Science Sealing Films for PCR and Storage

Detachable, peelable sealing films ideal for PCR, ELISA and sample storage.

215.00€

Specifications

Sterility Non-sterile
Autoclavable Not Autoclavable
Width (Metric) 9 cm
Pierceable No
Temperature Range (Metric) -80°C to +120°C
View More Specs

Products 2
Product Code Brand Length (Metric) Material Price Quantity & Availability  
Product Code Brand Length (Metric) Material Price Quantity & Availability  
17891242
View Documents
BRAND™
701376
17.2 cm Polyester
215.00€
Pack of 100
In Stock
to see stock.
Add to basket
 
17801252
View Documents
BRAND™
701377
16.5 cm Aluminum
215.00€
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

  • Due to its adhesive, film can be used down to -80°C
  • DMSO-resistant and resistant to common solvents
  • Can be removed without leaving any residue so that work can continue unhindered after transport or storage
Specifications

Specifications

Non-sterile
9 cm
-80°C to +120°C
Not Autoclavable
No
PCR Ready
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.