Learn More
Liebherr™ LFK Series Laboratory Refrigerator
Ensure maximum temperature stability. Liebherr™ LFK Series Laboratory Refrigerator is available in a range of sizes to meet every lab’s needs. Solid and glass door units feature automatic defrosting and rapid cooling. Electronic regulator includes audible and visual open door and high-temp alarms.
Brand:  Liebherr™ LFKUEX 1621
Product Code. 12527136
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Advanced laboratory coolers
  • Electronic temperature regulation for maximum temperature stability
  • Digital display control panel with audible and visual open door and high-temperature alarms
  • Rapid cooling after door opening with eco-friendly refrigerant
  • ''Anti-freezing'' safety thermostat and automatic defrosting with collection tray
Constructed for durable, efficient operation
  • Constructed with high-density polyurethane insulation
  • ABS tanks with ATEX secure tank options
  • Epoxy treated steel bodywork, with solid and glass front doors
  • Range of sizes, from table-top to large capacity units
  • Reversible doors with key lock for safe storage of valuable goods
Specifications
Specifications
141 L
Solid
Automatic
Epoxy-coated Steel
CE, ISO 14001, ISO 9001
Static
LFKUEX1621
43.5 cm
66.7 cm
45.5 cm
50 Hz
39 kg
2°C to 12°C
3
R600A
Polyurethane
White
ATEX secure tank 
61.5 cm
82 cm
60 cm
230 V
100 w