Learn More
Tocris™ Lenalidomide 4'-PEG3-amine
Cereblon ligand with PEG linker and terminal amine for onward chemistry
Brand:  Tocris™ 7076/25
182.29 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18727106
  • 232.00€ / 25mg
Explore more special offers
Description
Description
Lenalidomide 4'-PEG3-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG3 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
3-[4-[[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione dihydrochloride
Degrader Building Blocks
0.95
25 mg
C21H30N4O60.2 HCl