Learn More
Tocris Bioscience™ Lenalidomide 4'-PEG1-azide
Cereblon ligand with PEG linker and terminal azide for onward chemistry
Brand:  Tocris Bioscience™ 7513/25
273.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18717856
  • 347.00€ / 25mg
Explore more special offers
Description
Description
Lenalidomide 4'-PEG1-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
3-[4-[[2-(2-Azidoethoxy)ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione
25 mg
C17H20N6O4
2399455-45-7
Degrader Building Blocks
0.95