missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ LDPE Screw Caps for Series 301 and 302 Narrow Neck Bottles

For use with Series 301/302 3000mL capacity narrow-neck bottles. Also fit Series 303 wide-neck bottles with capacity of 50 and 100 mL.

Brand:  Kautex™ 2000090607

Product Code. 15981401

  • 49.14€ / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • LDPE construction
  • Natural color
  • Cap size OD: 32 mm
  • For use with Series 301/302 narrow neck bottles with capacity of 3000 mL
  • Also fit Series 303 wide neck bottles with capacity of 50 and 100 mL
  • TRUSTED_SUSTAINABILITY
Specifications

Specifications

Leakproof Closure
3000 mL Narrow Neck Bottles, Series 301 and 302; 50 and 100 mL Wide Neck Bottles, Series 303
32 mm
Natural color
LDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions