Learn More
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates Product Code.: 17830122
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
Product Code. 17830122
24-410 mm, LDPE Foam, 250
missing translation for 'orderingAttributeHoverText'
Closure Size:
24-410 mm
Liner Material:
LDPE Foam
No. per Case:
250
Product Code. 17830122
Brand: Thermo Scientific™ 0672410WBP
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these LDPE foam-lined caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ LDPE Foam-lined Polypropylene White Caps are for general-purpose HDPE bulk separate containers. The same high quality caps used on our Thermo Scientific certified HDPE environmental sample containers. Bottles sold separately (013-series and 012-series). Unprocessed and uncertified.
  • LDPE foam-lined white polypropylene cap
  • Bottles sold separately: 013-series and 012-series
  • Recommended for general purpose sampling where cleaning and certification is not required, or ready for your own cleaning and certification procedures
  • Economically bulk packed
Includes: Cap only
TRUSTED_SUSTAINABILITY
Specifications
Certification Parameters None
Color White
Description White PP Cap
Material Polypropylene
No. per Case 250
Septa N/A
Level Unprocessed
Liner Material LDPE Foam
Certifications/Compliance Manufactured under an ISO certified quality management system.
Closure Included Yes
Shape Round
Product Type Closure
Closure Liner LDPE Foam
Closure Size 24-410 mm
Show More Show Less
Product Content Correction
Product Title
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates >