Learn More
Semadeni™ LDPE Containers
Brand: Semadeni™ 875
Additional Details : Weight : 0.80000kg
Specifications
3.5 L | |
Natural Translucent | |
Rectangular | |
Snap-Cap | |
1 Each | |
208 mm | |
94 mm |
Snap Cap | |
Polyethylene | |
Packaging, Storage | |
Container | |
100 x 100 x 95 mm | |
208 mm |