missing translation for 'onlineSavingsMsg'
Learn More

Semadeni™ LDPE Containers

Brand:  Semadeni™ 871

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11842642

  • 6.05€ / Each
Estimated Shipment: 30-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1 L
Square
1 Each
100 mm
LDPE
Container
100 x 100 x 130 mm
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions