missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LDN 193189 dihydrochloride

Potent and selective ALK2 and ALK3 inhibitor; inhibits BMP4 signaling; promotes neural induction of hPSCs

Brand:  Tocris Bioscience™ 6053/50

868.22 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18787766

  • 1135.00€ / 50mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

LDN 193189 dihydrochloride is a potent and selective ALK2 and ALK3 inhibitor (IC50 values are 5 and 30 nM, respectively); inhibits BMP4-mediated Smad1/5/8 activation. Exhibits >200-fold selectivity for BMP signaling over TGF-β signaling. Also exhibits selectivity over AMPK, PDGFR and MAPK signaling. Promotes neural induction of hPSCs in combination with SB 431542 (Cat.No. 1614). Also induces differentiation of hPSCs into nociceptive sensory neurons in combination with SB 431542 (Cat.No. 1614), SU 5402 (Cat.No.3300), CHIR 99021 (Cat.No. 4423) and DAPT (Cat.No. 2634).

LDN 193189 synthesized to Ancillary Material Grade also available.

For more information about how LDN 193189 dihydrochloride may be used, see our protocols: Generation of β cells from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Accelerated Induction of Cortical Neurons from hiPSCs .

Specifications

Specifications

4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
Soluble to 50mM in water and to 20mM in DMSO
C25H22N60.2 HCl
Solid
Bioactive Small Molecule
1435934-00-1
50 mg
BMP and Other Activin Receptor Inhibitors
479.4
Orange
98%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.