Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
The LD series of cryogenic dewars are used to store and distribute small amounts of liquid nitrogen. They feature extremely low evaporation rates and are convenient to use for various applications.
Brand:  Taylor Wharton™ LD10
 View more versions of this product
Product Code. 11768519
  • 750.00€ / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Cryogenic Storage Dewar
10 L
597 mm
6.6 kg
2.6 gal.
23.5 in.
14.6 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products