Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
The LD series of cryogenic dewars are used to store and distribute small amounts of liquid nitrogen. They feature extremely low evaporation rates and are convenient to use for various applications.
Brand: Taylor Wharton™ LD5
This item is not returnable.
View return policy
Specifications
Cryogenic Storage Dewar | |
5 L | |
445 mm | |
3.1 kg |
1.3 gal. | |
17.5 in. | |
6.8 lb. |