missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth, Miller (Granulated)

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

Brand:  Fisher BioReagents™ BP9723-500

92.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11345992

  • 116.00€ / 500g
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Granulated Powder
Microbiology Media
For maintenance and propagation of Escherichia coli in molecular biology procedures
500 g
LB Broth
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.