missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ ERK2 A375 Cell Line

Product Code. 10461215
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10461215

Brand: Thermo Scientific™ K1798

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

The Map Kinase (MapK) signal transduction cascade is activated by growth factors such as EGF, PDGF, and HGF.

The Map Kinase (MapK) signal transduction cascade is activated by growth factors such as EGF, PDGF, and HGF. Binding of these factors to their respective cell surface receptors results in the initiation of receptor tyrosine kinase activity, which leads to the sequential phospho-activation of downstream kinases such as Ras, Raf, MEK, and Erk 1/2. Activated MEK phosphorylates Erk2 (Mapk1) proteins at a specific Thr-Tyr motif (Thr/Tyr 185/187). A number of constitutive active kinase mutants within the MAP kinase pathway have been implicated in oncogenesis. One of these mutants is BRAF(V600E), which is the most predominant oncogenic BRAF mutant. The human melanoma cell line A375 endogenously expresses BRAF(V600E), which leads to the constitutive activation of the MAP kinase pathway and phosphorylation of Erk2 in the absence of ligands.

LanthaScreenTM Erk2 A375 constitutively expresses GFP-Erk2 under control of a CMV promotor. Using this cell line, a homogenous immuno-assay has been developed in which the phosphorylation state of GFP-Erk2 is detected in cell lysates using a terbium-labeled anti-pTpY-185/187-Erk2 antibody, in a time-resolved FRET (TR-FRET) readout. This cell line can be used to evaluate compound activity against BRAF(V600E). GFP-Erk2 lentivirus was transduced into A375 cells, followed by selection with Blasticidin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as sorting marker. Using the lytic TR-FRET immuno-assay, this cell line is validated for EC50 and Z' under optimized conditions using EGF as a ligand for GFP-Erk2 phosphorylation. This assay has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance and cell lysis/equilibration time. Additional information using alternate stimuli and small molecule inhibitor is also provided.

TRUSTED_SUSTAINABILITY

Specifications

Antagonists Raf Inhibitor I
Assay Entry Cell-based terbium phospho-specific activity (stable)
Cell State Dividing Cells
Content And Storage LanthaScreen™ ERK2 A375 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) TR-FRET
Gene Symbol ERK2
Primary Agonist Constitutive
Product Line LanthaScreen
Quantity 1 Vial
Readout End Point
Target Entry ERK2
Type Cell Line
Cell Line A375
System Type LanthaScreen™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.