missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum for AAS, 2% La in HCl 2%, Honeywell Fluka™

Brand:  Honeywell Fluka E4400-100ML

Product Code. 16684454

  • 232.29€ / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lanthanum for AAS
1.02 g/cm3
100 mL
Standards and Modifiers for AAS
<2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions