Learn More
Brady™ Laminated Polyester Radioactive Warning Sign
4.00€ - 9.65€
Specifications
Material | Polyester |
---|---|
Quantity | 1 Ea. |
Adhesive Type | Permanent Acrylic |
Description | Laminated Polyester |
For Use With (Application) | Used for indoor and outdoor safety signs |
Specifications
Polyester | |
Permanent Acrylic | |
Used for indoor and outdoor safety signs | |
Warning of Radioactive materials or radiation ioniserenden | |
Black on Yellow |
1 Ea. | |
Laminated Polyester | |
Triangular | |
Radioactive Warning Sign |