Learn More
Dostmann Elecronic Labtherm Insertion Thermometer
With a measuring range of 39.9°C to 249.9°C and an accuracy of ±0.5°C, this LabTherm digital catering thermometer is particularly suitable to measure liquids, food, etc.
Brand:  Dostmann Elecronic 5020-0398
Additional Details : Weight : 0.07200kg
Product Code. 10526795
  • 40.05€ / Each
In Stock
to see stock.
Récupel-bijdrage uitgesloten
Description
Description
  • Labtherm Insertion thermometer
  • Ergonomic shaped pocket thermometer with robust inser­tion tip
  • The long sensor tube is particular suitable to measure in liquids, food etc
  • LabTherm digital catering thermometer with a precise read-out of temperature over the range of -39.9°C to 249.9°C 
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Services