Learn More
Koettermann™ Laboratory Swivel Chair with Wheels, Adjustable Height
Koettermann™ Laboratory Swivel Chair with Wheels, Adjustable Height features casters and an adjustable height range.
Brand: Koettermann™ 942-00001
480.60 EUR valid until 2024-11-01
Use promo code "21705" to get your promotional price.
This item is not returnable.
View return policy
Description
- Swivel chair
- Casters
- Adjustable height
Specifications
Lab Swivel Chair | |
Adjustable Height | |
Casters |
Black | |
420 to 540 mm | |
Yes |