Learn More
Buerkle™ LaboPlast™ Micro Spatulas
Use to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids. Buerkle™ LaboPlast™ Micro Spatulas facilitates precise work with sturdy centre handle.
Brand: Buerkle™ 5378-0032
31.68 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Double scoop spatula with rounded ends
- Sturdy center handle facilitates precise work
- Large package (100 pieces)
Specifications
Polystyrene | |
Non-sterile | |
Bulk pack, 100 pieces |
1 mL, 0.25 mL | |
EU food regulations |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Buerkle LaboPlast Micro Spatulas
Topsellers prelevement pharmaceutique
File size: 612.9 kB
Topsellers pharma sampling
File size: 611.7 kB
Topsellers disposamplers
File size: 1.3 MB
Topsellers disposamplers
File size: 1.3 MB
Brochure LaboPlast SteriPlast scoops spoons spatulas
File size: 608.8 kB
Brochure LaboPlast SteriPlast pelles cuilleres spatules
File size: 610.2 kB
Product brochure Spoon spatula Micro spatula
File size: 66.3 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
251174 | Certificate of Compliance | 18/03/2025 | 5378-0032 |
244558 | Certificate of Compliance | 19/11/2024 | 5378-0032 |
243244 | Certificate of Compliance | 16/08/2024 | 5378-0032 |
240857 | Certificate of Compliance | 12/03/2024 | 5378-0032 |
233149 | Certificate of Compliance | 25/07/2023 | 5378-0032 |
Product Content Correction
Product Title
Buerkle™ LaboPlast™ Micro Spatulas > 1 mL/0.25 mL