Learn More
Hirschmann™ Labopette™ Variable Volume Single Channel Manual Pipette
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand: Hirschmann™ 9475405
This item is not returnable.
View return policy
Description
- Variable volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
0.8% | |
100 to 5000 μL | |
Clear | |
Labopette™ | |
500 to 5000 μL | |
10 μL |
CE, IVD 98/79 EC | |
Autoclavable | |
1 | |
Labopettor | |
Calibration tool and pipette holder, individual serial number and calibration certificate |