Learn More
Labconco™ Labconco Soda Acid Trap Insert for Pre-2017 Trap
Product Code. 15103095 Shop All Labconco Corporation Products
Click to view available options
:
Acid Trap Insert
Product Code. 15103095
Brand: Labconco™ 7772100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Traps are connected between the evaporator and vacuum pump to provide additional protection to the pump.
Specifications
Product Type Soda Acid Trap Insert
Weight (English) 4.0 lb.
Weight (Metric) 1.8 kg