Learn More
Labconco™ Glass Trap Insert for CentriVap™ System
Protects CentriVap Cold Trap reservoir and lid from corrosives.
Brand:  Labconco™ 7397605
Product Code. 10492960
  • 1115.00€ / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Protects CentriVap Cold Trap reservoir and lid from corrosives.
  • Included with CentriVap Acid Systems; accessory for other systems
  • Not compatible with pre-2005 Cold Traps
Specifications
Specifications
Trap Insert
CentriVap™ concentrator cold trap
1.8 kg
2550 mL
4 lb.