missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ AtmosPure™ Re-Gen Gas Purifiers

Automatic regeneration process completed in as little as 13 hours while unattended

Brand:  Labconco™ 5121835

 View more versions of this product

Product Code. 15937590

  • 16005.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Produces an ultrapure inert atmosphere for materials sensitive to moisture and oxygen
  • Has capacity to remove up to five liters of oxygen (13.500 ppm) and 660 grams of moisture (18 million ppm) at standard temperature and pressure
  • Durable powder-coated steel housing
  • Three microprocessor-controlled solenoid valves, located on the back
  • Flow control valve sets 4% hydrogen between 10-25 CFH
  • Regeneration heater delivers 180 watts for 5 hours
  • Blower button toggles circulation air purifier blower between ON, OFF and AUTO
  • Designed to fit on the accessory Glove Box Base Stand beneath the transfer chamber

  • Cylinder contains 5 pounds of copper catalyst and 5 pounds of molecular sieve
  • Automatically compensates for power outages and starts the regeneration process over if interrupted for 10 minutes
  • Has internal stainless steel gas purification lines and communications port on the back
  • 0.75 in. inlet/outlet compression fittings for connection to protector fiberglass or atmosphere glove boxes

Specifications

Specifications

0.75 in. Fitting
Controlled atmosphere glove boxes
Stainless Steel
14.9 in.
21.9 in.
208-230V, 50/60Hz, 10A
Re-Gen Gas Purifier
Epoxy-coated Steel
Schuko
38 cm
56 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions