Learn More
Prohibitin (Mitochondrial Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Prohibitin (Mitochondrial Marker) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-261-B0
Description
Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin appears to be a very reliable marker of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization, and tumor suppression.Host Species: Mouse
Clone: II-14-10
Isotype: IgG1
Species Reactivity: Human, Pig, Rat, Mouse, and Chicken. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant rat prohibitin protein
Molecular Weight: 30kDa
Positive Control: MAD109 or PC12 cells, Heart
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Prohibitin (Mitochondrial Marker) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Apoptosis | |
Chicken, Human, Mouse, Porcine, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
II-14-10 | |
Biotin | |
Purified recombinant rat prohibitin protein | |
RUO | |
Primary | |
Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.