Learn More
pan-Actin Ab-5 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.
Brand: Epredia MS-1295-P
Description
Host Species: Mouse
Clone: ACTN05; same as C4
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Mouse, Dog, Pig, Rabbit, Rat, Chicken, Dictyostelium discoideum and Physarum polycephalum. Others not known.
Epitope: Not determined
Immunogen: Chicken gizzard actin
Positive Control: HeLa cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Actin, pan Ab-5 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Muscle Biology | |
Bovine, Canine, Chicken, Human, Mouse, P. polycephalum, Porcine, Rabbit, Rat | |
IgG1 κ |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
ACTN05; same as C4 | |
Unconjugated | |
Chicken gizzard actin | |
RUO | |
Primary | |
Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.