missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals
57.70€ - 190.00€
Chemical Identifiers
CAS | 6306-52-1 |
---|---|
Molecular Formula | C6H13ClNO2 |
Molecular Weight (g/mol) | 166.63 |
MDL Number | MFCD00012497 |
InChI Key | RFMIEHOYULVMNZ-UHFFFAOYNA-N |
Synonym | l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride |
PubChem CID | 111190 |
IUPAC Name | methyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
SMILES | [Cl].COC(=O)C(N)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402618
|
Thermo Scientific Alfa Aesar
A15892.09 |
10 g |
57.70€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412618
|
Thermo Scientific Alfa Aesar
A15892.18 |
50 g |
190.00€
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6306-52-1 | |
166.63 | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
111190 | |
[Cl].COC(=O)C(N)C(C)C |
C6H13ClNO2 | |
MFCD00012497 | |
l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride | |
methyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
Specifications
166°C (decomposition) | |
C6H13ClNO2 | |
3594960 | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
methyl (2S)-2-amino-3-methylbutanoate;hydrochloride | |
111190 | |
99% | |
L-Valine methyl ester hydrochloride |
6306-52-1 | |
MFCD00012497 | |
l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride | |
[Cl].COC(=O)C(N)C(C)C | |
166.63 | |
167.64 | |
+16° (c=2 in Water) |
Safety and Handling
missing translation for 'einecsNumber' : 228-620-9
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only