missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
19.70€ - 54.90€
Chemical Identifiers
CAS | 17609-47-1 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00012511 |
InChI Key | PQGVTLQEKCJXKF-RGMNGODLSA-N |
Synonym | l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl |
PubChem CID | 87181 |
SMILES | Cl.CCOC(=O)[C@@H](N)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465798
|
Thermo Scientific Alfa Aesar
H59466.03 |
1 g |
19.70€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475798
|
Thermo Scientific Alfa Aesar
H59466.06 |
5 g |
54.90€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17609-47-1 | |
181.66 | |
PQGVTLQEKCJXKF-RGMNGODLSA-N | |
87181 |
C7H16ClNO2 | |
MFCD00012511 | |
l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
Cl.CCOC(=O)[C@@H](N)C(C)C |
Specifications
102°C to 105°C | |
C7H16ClNO2 | |
MFCD00012511 | |
PQGVTLQEKCJXKF-RGMNGODLSA-N | |
ethyl (2S)-2-amino-3-methylbutanoate;hydrochloride | |
87181 | |
98% | |
L-Valine ethyl ester hydrochloride |
17609-47-1 | |
(CH3)2CHCH(NH2)CO2C2H5·HCl | |
l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
Cl.CCOC(=O)[C@@H](N)C(C)C | |
181.66 | |
181.66 | |
+6.7° (c=2 in H2O) |
Safety and Handling
missing translation for 'einecsNumber' : 241-580-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only