missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-threonine methyleester hydrochloride, +98%, Thermo Scientific Chemicals
41.00€ - 504.00€
Chemical Identifiers
CAS | 39994-75-7 |
---|---|
Molecular Formula | C5H12ClNO3 |
Molecular Weight (g/mol) | 169.61 |
MDL Number | MFCD00037677 |
InChI Key | OZSJLLVVZFTDEY-HJXLNUONSA-N |
Synonym | l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl |
PubChem CID | 2734893 |
IUPAC Name | methyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride |
SMILES | Cl.COC(=O)[C@@H](N)[C@@H](C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10482083
|
Thermo Scientific Acros
376690010 |
1 g | Glass Bottle |
41.00€
1g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10547113
|
Thermo Scientific Acros
376690050 |
5 g | Glass Bottle |
137.00€
5g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10500174
|
Thermo Scientific Acros
376690250 |
25 g | Glass Bottle |
504.00€
25g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39994-75-7 | |
169.61 | |
OZSJLLVVZFTDEY-HJXLNUONSA-N | |
2734893 | |
Cl.COC(=O)[C@@H](N)[C@@H](C)O |
C5H12ClNO3 | |
MFCD00037677 | |
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl | |
methyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride |
Specifications
39994-75-7 | |
98+% | |
Authentic | |
MFCD00037677 | |
OZSJLLVVZFTDEY-HJXLNUONSA-N | |
methyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride | |
169.61 | |
169.61 | |
1% Max. | |
Powder |
White | |
C5H12ClNO3 | |
CH3CH(OH)CH(NH2)C(O)OCH3·HCl | |
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl | |
Cl.COC(=O)[C@@H](N)[C@@H](C)O | |
-9.5° to -10.5° (20°C, 589nm) (c=1, Methanol, 20°C) | |
2734893 | |
≥98.0% (on dry substance) | |
−10° (20°C c=1,methanol) | |
L-Threonine methyleester hydrochloride |
RUO – Research Use Only