Learn More
Copan Diagnostics L-Shaped Spreaders
L-shaped spreaders
Brand: Copan Diagnostics 174CS05
Description
L-shaped spreaders
L-Shaped Plastic Spreaders
- For spreading liquid samples
- Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
- Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
- Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Culture Plates | |
L-Shape | |
5/peel pack; 1000 spreaders/box | |
Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |
5/peel pk.; 1000 Spreaders/Box | |
Plastic | |
Sterile |