missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Serine methyleester hydrochloride, 98%
41.70€ - 372.00€
Chemical Identifiers
CAS | 5680-80-8 |
---|---|
Molecular Formula | C4H10ClNO3 |
Molecular Weight (g/mol) | 155.578 |
MDL Number | MFCD00063680 |
InChI Key | NDBQJIBNNUJNHA-DFWYDOINSA-N |
Synonym | l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl |
PubChem CID | 2723730 |
IUPAC Name | methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride |
SMILES | COC(=O)C(CO)N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091430
|
Thermo Scientific Chemicals
132670050 |
5 g | Glass Bottle |
41.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10513111
|
Thermo Scientific Chemicals
132670250 |
25 g | Glass Bottle |
115.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540584
|
Thermo Scientific Chemicals
132671000 |
100 g | Glass Bottle |
372.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5680-80-8 | |
155.578 | |
NDBQJIBNNUJNHA-DFWYDOINSA-N | |
2723730 | |
COC(=O)C(CO)N.Cl |
C4H10ClNO3 | |
MFCD00063680 | |
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl | |
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride |
Specifications
160.0°C to 165.0°C | |
White | |
C4H10ClNO3 | |
HOCH2CH(NH2)CO2CH3·HCl | |
04, 506 | |
Solubility in water: soluble. Other solubilities: soluble in methanol | |
COC(=O)C(CO)N.Cl | |
+ 4.50 | |
2723730 | |
98% | |
1% Max. (K.F.) | |
Crystalline Powder or Crystals |
5680-80-8 | |
98% | |
Authentic | |
MFCD00063680 | |
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl | |
NDBQJIBNNUJNHA-DFWYDOINSA-N | |
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride | |
155.578 | |
155.58 | |
0.1% max. | |
+ 4.50 (20.00°C c=2,methanol) | |
L-Serine methyl ester hydrochloride |
RUO – Research Use Only