Learn More
Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent
99.40€ - 331.00€
Chemical Identifiers
CAS | 63-91-2 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00064227 |
InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
Synonym | l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh |
PubChem CID | 6140 |
ChEBI | CHEBI:17295 |
SMILES | NC(CC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496079
|
Thermo Scientific Chemicals
J63925.22 |
100 g |
99.40€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406089
|
Thermo Scientific Chemicals
J63925.30 |
250 g |
185.00€
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416089
|
Thermo Scientific Chemicals
J63925.36 |
500 g |
331.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.
Solubility
Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH{4}OH
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
63-91-2 | |
165.19 | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
6140 | |
NC(CC1=CC=CC=C1)C(O)=O |
C9H11NO2 | |
MFCD00064227 | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
CHEBI:17295 |
Specifications
63-91-2 | |
C9H11NO2 | |
1910408 | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
2-amino-3-phenylpropanoic acid | |
6140 | |
165.19 | |
Powder |
White | |
MFCD00064227 | |
14,7271 | |
Soluble in water; Aqueous solubility increases with warming; Also soluble in methanol,ethanol or 1M NH4OH | |
NC(CC1=CC=CC=C1)C(O)=O | |
165.19 | |
CHEBI:17295 | |
Cell Culture Reagent | |
L-Phenylalanine |
Safety and Handling
missing translation for 'einecsNumber' : 200-568-1
missing translation for 'rtecsNumber' : AY7535000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only