missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoxide

42.20€ - 646.00€

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00063093
InChI Key QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Synonym l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
PubChem CID 158980
ChEBI CHEBI:17016
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405179
View Documents Promotion Details
Thermo Scientific Chemicals
J62873.03
1 g
42.20€
1g
Please to purchase this item. Need a web account? Register with us today!
15415179
View Documents Promotion Details
Thermo Scientific Chemicals
J62873.06
5 g
160.00€
5g
Please to purchase this item. Need a web account? Register with us today!
15425179
View Documents Promotion Details
Thermo Scientific Chemicals
J62873.14
25 g
646.00€
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3226-65-1
165.21
QEFRNWWLZKMPFJ-YGVKFDHGSA-N
158980
(2S)-2-amino-4-methylsulfinylbutanoic acid
C5H11NO3S
MFCD00063093
l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
CHEBI:17016
CS(=O)CC[C@H](N)C(O)=O

Specifications

3226-65-1
C5H11NO3S
4129939
QEFRNWWLZKMPFJ-YGVKFDHGSA-N
(2S)-2-amino-4-methylsulfinylbutanoic acid
158980
165.21
L-Methionine sulfoxide
White
MFCD00063093
l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
CS(=O)CC[C@H](N)C(O)=O
165.21
CHEBI:17016
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 221-758-0

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only