Learn More
Gibco™ L-Methionine (Methyl-13C)
L-Methionine is a common sulfur-containing amino acid that is commonly used for the isotopic labeling of methionine protein residues for NMR spectroscopy studies.
Brand:  Gibco™ A39248
Product Code. 16525651
  • 239.00€ / 225mg
Explore more special offers
Description
Description
Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.
EC Number: 200-562-9 (unlabeled)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
(S)-2-Amino-4-(Methylmercapto)butyric acid / L-2-Amino-4-(methylthio)cutanoic acid
225 mg
Powder
150.2
>98%
Wet Ice