missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
54.60€ - 750.00€
Chemical Identifiers
CAS | 7517-19-3 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00012494 |
InChI Key | DODCBMODXGJOKD-HQFIOVGONA-N |
Synonym | l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride |
PubChem CID | 2723695 |
IUPAC Name | methyl (2S)-2-amino-4-methylpentanoate;hydrochloride |
SMILES | Cl.COC(=O)[C@@H](N)CC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11492697
|
Thermo Scientific Alfa Aesar
A14897.14 |
25 g |
54.60€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402707
|
Thermo Scientific Alfa Aesar
A14897.22 |
100 g |
162.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412707
|
Thermo Scientific Alfa Aesar
A14897.36 |
500 g |
750.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7517-19-3 | |
181.66 | |
DODCBMODXGJOKD-HQFIOVGONA-N | |
2723695 | |
Cl.COC(=O)[C@@H](N)CC(C)C |
C7H16ClNO2 | |
MFCD00012494 | |
l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride | |
methyl (2S)-2-amino-4-methylpentanoate;hydrochloride |
Specifications
149°C to 151°C | |
C7H16ClNO2 | |
3595133 | |
DODCBMODXGJOKD-HQFIOVGONA-N | |
methyl (2S)-2-amino-4-methylpentanoate;hydrochloride | |
2723695 | |
99% | |
Hygroscopic |
7517-19-3 | |
MFCD00012494 | |
l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride | |
Cl.COC(=O)[C@@H](N)CC(C)C | |
181.66 | |
181.66 | |
+13° (c=2 in Water) | |
L-Leucine methyl ester hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 231-375-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only