Learn More
L-Histidine monohydrochloride monohydrate, Cell Culture Reagent, Thermo Scientific Chemicals
19.40€ - 348.00€
Chemical Identifiers
CAS | 5934-29-2 |
---|---|
Molecular Formula | C6H12ClN3O3 |
Molecular Weight (g/mol) | 209.63 |
MDL Number | MFCD00151027 |
InChI Key | CMXXUDSWGMGYLZ-XRIGFGBMSA-N |
Synonym | l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate |
PubChem CID | 165377 |
IUPAC Name | (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
SMILES | C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15400197
|
Thermo Scientific Alfa Aesar
J61465.09 |
10 g |
19.40€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410197
|
Thermo Scientific Alfa Aesar
J61465.22 |
100 g |
97.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420197
|
Thermo Scientific Alfa Aesar
J61465.36 |
500 g |
232.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430197
|
Thermo Scientific Alfa Aesar
J61465.A1 |
1 kg | N/A | ||||||
Description
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.
Solubility
Freely soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
5934-29-2 | |
209.63 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
165377 | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
C6H12ClN3O3 | |
MFCD00151027 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
Specifications
∽240°C (decomposition) | |
White | |
MFCD00151027 | |
14,4720 | |
Freely soluble in water | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl | |
209.63 | |
209.63 (191.62 Anhydrous) | |
+10° (c=5 in 5N HCl) | |
L-Histidine monohydrochloride monohydrate |
5934-29-2 | |
C6H12ClN3O3 | |
4168261 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride | |
165377 | |
Cell Culture Reagent | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 211-438-9
missing translation for 'rtecsNumber' : MS3119000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only