Learn More
Thermo Scientific Chemicals L-Glutamic acid hydrochloride, 99%
38.50€ - 108.00€
Chemical Identifiers
CAS | 138-15-8 |
---|---|
Molecular Formula | C5H10ClNO4 |
Molecular Weight (g/mol) | 183.588 |
MDL Number | MFCD00012619 |
InChI Key | RPAJSBKBKSSMLJ-DFWYDOINSA-N |
Synonym | l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin |
PubChem CID | 2723891 |
IUPAC Name | (2S)-2-aminopentanedioic acid;hydrochloride |
SMILES | C(CC(=O)O)C(C(=O)O)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424154
|
Thermo Scientific Chemicals
A12505.30 |
250 g |
38.50€
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414154
|
Thermo Scientific Chemicals
A12505.0B |
1000 g |
108.00€
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.
Solubility
Soluble in water (490 g/L).
Notes
Hygroscopic. Incompatible with oxidizing agents.
Chemical Identifiers
138-15-8 | |
183.588 | |
RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
2723891 | |
C(CC(=O)O)C(C(=O)O)N.Cl |
C5H10ClNO4 | |
MFCD00012619 | |
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
(2S)-2-aminopentanedioic acid;hydrochloride |
Specifications
203°C (decomposition) | |
C5H10ClNO4 | |
3565569 | |
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
(2S)-2-aminopentanedioic acid;hydrochloride | |
2723891 | |
99% | |
+23° (c=6 in Water) | |
L-Glutamic acid hydrochloride |
138-15-8 | |
MFCD00012619 | |
14,4469 | |
Soluble in water (490g/L). | |
C(CC(=O)O)C(C(=O)O)N.Cl | |
183.588 | |
183.59 | |
1.525 | |
Hygroscopic |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'einecsNumber' : 205-315-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only