Learn More
Thermo Scientific Chemicals L(+)-Glutamic acid hydrochloride, 99%
20.50€ - 219.00€
Chemical Identifiers
CAS | 138-15-8 |
---|---|
Molecular Formula | C5H10ClNO4 |
Molecular Weight (g/mol) | 183.588 |
MDL Number | MFCD00012619 |
InChI Key | RPAJSBKBKSSMLJ-DFWYDOINSA-N |
Synonym | l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin |
PubChem CID | 2723891 |
IUPAC Name | (2S)-2-aminopentanedioic acid;hydrochloride |
SMILES | C(CC(=O)O)C(C(=O)O)N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286210
|
Thermo Scientific Chemicals
165231000 |
100 g | Glass Bottle |
20.50€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091860
|
Thermo Scientific Chemicals
165235000 |
500 g | Glass Bottle |
62.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137943
|
Thermo Scientific Chemicals
165230025 |
2.5 kg | Plastic Bottle |
219.00€
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138-15-8 | |
183.588 | |
RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
2723891 | |
C(CC(=O)O)C(C(=O)O)N.Cl |
C5H10ClNO4 | |
MFCD00012619 | |
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
(2S)-2-aminopentanedioic acid;hydrochloride |
Specifications
200ppm max. | |
1ppm max. | |
White | |
99% | |
Authentic | |
10ppm max. | |
04, 488 | |
15, 4505 | |
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
C(CC(=O)O)C(C(=O)O)N.Cl | |
+25.2° to +25.8° (20°C, 589nm) (c=10, 2N HCl) | |
2723891 | |
183.59 | |
98.5 to 101.5% | |
+25.50° (20°C c=10,2N HCl) | |
L(+)-Glutamic acid hydrochloride, 99% |
138-15-8 | |
1 to 2.5 (2.5% soln. at 25°C) | |
10ppm max. | |
C5H10ClNO4 | |
HO2CCH2CH2CH(NH2)CO2H·HCl | |
MFCD00012619 | |
0.5% max. (105°C, 5 hrs) | |
0.5% max. (TLC) | |
RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
(2S)-2-aminopentanedioic acid;hydrochloride | |
183.588 | |
200ppm max. | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
0.1% max. | |
Fine Crystalline Powder or Crystals |
Safety and Handling
GHS P Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
missing translation for 'einecsNumber' : 205-315-9
missing translation for 'tsca' : TSCA
- L(+)-Glutamic acid hydrochloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only