missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
74.60€ - 673.00€
Chemical Identifiers
CAS | 56-86-0 |
---|---|
Molecular Formula | C5H9NO4 |
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00002634 |
InChI Key | WHUUTDBJXJRKMK-VKHMYHEASA-N |
Synonym | l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex |
PubChem CID | 33032 |
ChEBI | CHEBI:16015 |
IUPAC Name | (2 S)-2-aminopentanedioic acid |
SMILES | C(CC(=O)O)C(C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15845288
|
Thermo Scientific
J16215.A1 |
1 kg | Plastic Bottle |
74.60€
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15855288
|
Thermo Scientific
J16215.A7 |
5 kg | Plastic Bottle |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
15865288
|
Thermo Scientific
J16215.C4 |
25 kg | Plastic Bottle |
673.00€
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
56-86-0 | |
147.13 | |
WHUUTDBJXJRKMK-VKHMYHEASA-N | |
33032 | |
(2 S)-2-aminopentanedioic acid |
C5H9NO4 | |
MFCD00002634 | |
l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
CHEBI:16015 | |
C(CC(=O)O)C(C(=O)O)N |
Specifications
189°C to 192°C (decomposition) | |
C5H9NO4 | |
MFCD00002634 | |
l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
C(CC(=O)O)C(C(=O)O)N | |
147.13 | |
CHEBI:16015 | |
98.5 to 101.5% | |
1.525 | |
Dry basis |
56-86-0 | |
HOOCCH2CH2CH(NH2)COOH | |
1723801 | |
WHUUTDBJXJRKMK-VKHMYHEASA-N | |
(2 S)-2-aminopentanedioic acid | |
33032 | |
147.13 | |
1.525 | |
Crystalline Powder | |
L-Glutamic acid |
Safety and Handling
H500
missing translation for 'einecsNumber' : 200-293-7
missing translation for 'rtecsNumber' : LZ9700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only