missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid 5-benzyl ester, 99%, Thermo Scientific Chemicals
25.40€ - 225.00€
Chemical Identifiers
CAS | 1676-73-9 |
---|---|
Molecular Formula | C12H15NO4 |
Molecular Weight (g/mol) | 237.255 |
MDL Number | MFCD00002633 |
InChI Key | BGGHCRNCRWQABU-JTQLQIEISA-N |
Synonym | h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl |
PubChem CID | 122337 |
IUPAC Name | (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid |
SMILES | C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429727
|
Thermo Scientific Alfa Aesar
B22184.03 |
1 g |
25.40€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439727
|
Thermo Scientific Alfa Aesar
B22184.06 |
5 g |
80.30€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449727
|
Thermo Scientific Alfa Aesar
B22184.14 |
25 g |
225.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1676-73-9 | |
237.255 | |
BGGHCRNCRWQABU-JTQLQIEISA-N | |
122337 | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N |
C12H15NO4 | |
MFCD00002633 | |
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl | |
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid |
Specifications
162°C to 165°C | |
C12H15NO4 | |
1885646 | |
BGGHCRNCRWQABU-JTQLQIEISA-N | |
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid | |
122337 | |
99% | |
L-Glutamic acid 5-benzyl ester |
1676-73-9 | |
MFCD00002633 | |
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N | |
237.255 | |
237.26 | |
+23° (c=1 in Acetic Acid) |
Safety and Handling
missing translation for 'einecsNumber' : 216-826-1
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only