missing translation for 'onlineSavingsMsg'
Learn More

L-CCG-l, Tocris Bioscience™

Product Code. 30001079
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent group II mGlu agonist

Potent group II metabotropic glutamate receptor agonist. More active than glutamate or (±)-trans-ACPD (Cat. No. 0187).

Specifications

CAS 117857-93-9
Molecular Formula C6H9NO4
Synonym 2s,1's,2's-2-carboxycyclopropyl glycine, l-ccg-l, l-ccg-i, ccpg, l-ccg-1, 3,4-cyclopropylglutamate, l-2-carboxypropyl glycine, 1s,2s-2-1s-1-amino-2-hydroxy-2-oxoethyl cyclopropane-1-carboxylic acid, l-ccgi, d0z7cz
InChI Key GZOVEPYOCJWRFC-HZLVTQRSSA-N
SMILES C1C(C1C(=O)O)C(C(=O)O)N
IUPAC Name (1S,2S)-2-[(S)-amino(carboxy)methyl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 159.141
PubChem CID 5310956
Formula Weight 159.14
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material L-CCG-l
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'