Learn More
Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent
55.90€ - 370.00€
Chemical Identifiers
CAS | 56-41-7 |
---|---|
Molecular Formula | C3H7NO2 |
Molecular Weight (g/mol) | 89.09 |
MDL Number | MFCD00064410 |
InChI Key | QNAYBMKLOCPYGJ-REOHCLBHSA-N |
Synonym | l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid |
PubChem CID | 5950 |
ChEBI | CHEBI:16977 |
IUPAC Name | (2S)-2-aminopropanoic acid |
SMILES | C[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463069
|
Thermo Scientific Chemicals
J60279.18 |
50 g |
55.90€
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473069
|
Thermo Scientific Chemicals
J60279.22 |
100 g |
100.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483069
|
Thermo Scientific Chemicals
J60279.36 |
500 g |
370.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.
Solubility
Soluble in water 166.5 g/L..
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Chemical Identifiers
56-41-7 | |
89.09 | |
QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
5950 | |
(2S)-2-aminopropanoic acid |
C3H7NO2 | |
MFCD00064410 | |
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
CHEBI:16977 | |
C[C@H](N)C(O)=O |
Specifications
∽295°C (decomposition) | |
White | |
MFCD00064410 | |
14,204 | |
Soluble in water 166.5g/L.. | |
C[C@H](N)C(O)=O | |
89.09 | |
CHEBI:16977 | |
Cell Culture Reagent | |
Powder |
56-41-7 | |
C3H7NO2 | |
1720248 | |
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
(2S)-2-aminopropanoic acid | |
5950 | |
89.09 | |
+14° (c=5 in 5N HCl) | |
L-Alanine |
Safety and Handling
missing translation for 'einecsNumber' : 200-273-8
missing translation for 'rtecsNumber' : AY2990000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only