missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L 012 sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Luminal-based chemiluminescent probe. Detects NAPDH oxidase-derived reactive oxygen and nitrogen species (ROS and RNS). Active in vitro and in vivo.
Specifications
Specifications
CAS | 143556-24-5 |
Molecular Formula | C13H9ClN4NaO2 |
Quantity | 10 mg |
Synonym | l 012 sodium salt |
InChI Key | WRVLDBOUZVBBCX-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)C2=C(C3=C(C(=O)NNC3=O)C(=N2)Cl)N.[Na] |
IUPAC Name | 8-amino-5-chloro-7-phenyl-2,3-dihydropyrido[3,4-d]pyridazine-1,4-dione;sodium |
Molecular Weight (g/mol) | 311.681 |
PubChem CID | 131848063 |
Formula Weight | 310.67 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction