Learn More
KU70 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10723-1-AP-150UL
 View more versions of this product
Product Code. 16827293
  • 410.00€ / 150µL
Description
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.
Specifications
Specifications
KU70
Polyclonal
Unconjugated
XRCC6
5 dRP lyase Ku70, 70 kDa subunit of Ku antigen, CTC75, CTCBF, DNA repair protein XRCC6, G22P1, KU70, ML8, Thyroid lupus autoantigen, TLAA, XRCC6
Rabbit
Antigen Affinity Chromatography
RUO
25019, 2547
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12956
Xrcc6
KU70 Fusion Protein Ag1043
150 μL
Primary
Human, Rat