Learn More
DWK Life Sciences Kontes™ Ultra-Ware™ Microfiltration Assembly with Stainless Steel Support and Ground Joint (47mm)
Recommended when filtering viscous or proteinaceous solutions to give the maximum flow rate
Brand: DWK Life Sciences 953855-1047
This item is not returnable.
View return policy
Description
Recommended when filtering viscous or proteinaceous solutions to give the maximum flow rate
- This apparatus is also used to produce ultra-clean filtrate since the stainless steel screen will not shed particles into the filtrate
- The ground joint connection eliminates the possible phthalate contamination that can occur when using silicone or neoprene stoppers
- Each apparatus is supplied with a funnel, an anodized aluminum clamp, a stainless steel support screen, a PTFE gasket, a 47mm glass support base and a filtration flask
- 80 x 80 304 stainless steel mesh, 0.0055 in. opening size, 19.4% open area, 0.007 in. wire diameter
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Microfiltration Assembly | |
Microfiltration Assembly | |
5.37 in. | |
17 in. |
Stainless Steel (316) | |
1 L | |
11.7 cm | |
43 cm |