Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
Brand:  DWK Life Sciences 953835-0000
1377.50 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
 View more versions of this product
Product Code. 11868522
  • 1450.00€ / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
  • The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microfiltration Assembly
Glass
500 mL (Funnel), 2000 mL (Flask)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products