Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
Brand: DWK Life Sciences 953845-0000
1548.50 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
- The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
- The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
- Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
- Joints are 40/35
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Microfiltration Assembly | |
Glass | |
Microfiltration Assembly | |
1000 mL (Funnel), 4000 mL (Flask) |