Learn More
DWK Life Sciences Kimble™ Kontes™ Cuvette Tube Washer for NMR Utility Washers
Replacement fask for utility washers.
Brand:  DWK Life Sciences 459961-0000
Product Code. 11316220
  • 303.00€ / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Utility washers are used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items
  • Unit is supplied with one gasket, a tube washer, an Erlenmeyer flask and a 3/8 in. hose connection
  • Replacement flask, 1 EA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cuvette Washer
For NMR utility washer
Cuvette Washer