Learn More
Kontes™ Membrane Filtration Glassware from Restek
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Brand:  Kimble Chase 953826-0000
 View more versions of this product
Product Code. 11739055
  • 646.00€ / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Support, 40/35 Joint
Microfiltration Apparatus
Fritted Glass
47 mm