Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Polyethylene Disc
Product Code. p-4917475
Click to view available options
Type:
Disc 30 to 50 μm
Disc, 20 μm
Unit Size:
Pack of 100
Product Code. 10749175
Brand: DWK Life Sciences 4201620020
Only null left
Add to basket
Add to basket
For Disposaflex Chromatography Columns
  • Sold 100/pack.
TRUSTED_SUSTAINABILITY
Specifications
Type Disc, 20 μm
For Use With Disposaflex Chromatography Column
Material Polyethylene