Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Packing Reservoir, with Joints
These heavy duty reservoirs are designed for use with 420510 and 420550 series chromatography columns with and without fritted glass supports or in combination with 420575 series flow control adapters.
Brand:  DWK Life Sciences 420570-0124
 View more versions of this product
Product Code. 13491239
  • 174.00€ / Each
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reservoirs are available in a variety of sizes and joint configurations
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Packing Reservoir; w/Joints; 100mL
Packing Reservoir, with Joints