Learn More
DWK Life Sciences Kimble™ Kontes™ Chromaflex Chromatography Fittings : Adapter with 1/2-20 to 1/4-28 Thread
Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads.
Brand:  DWK Life Sciences 420804-0001
Product Code. 11833063
  • 117.00€ / Pack of 2
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads.
  • CTFE Threaded Adapter with 1/2-20 to 1/4-28 Thread
  • Converts 1/2-20 threads on CHROMAFLEX™ column end fittings to 1/4-28 threads
  • Case of 2
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
1
Glass
Chromatography
Kimble Kontes Hi-Vac PTFE plug